Ammonia solution 25 - 30% (NH₃ basis) in water, Puriss., meets analytical specification of Ph.Eur., Fluka™
: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
:1336-21-6
:7664-41-7
:NH₃
:MFCD00066650
:37.7 °C (1.013 hPa)
:–92 °C
:0.90 g/cm³ (20 °C)
:35.05 g/mol
:Ambient
:2672
:8 III
:GHS07
:GHS05
- Environmentally Preferable:
- Assay:25.0 - 30.0 %
- Density (D 20/20):0.892 - 0.910
- Non-volatile matter:Max. 0.0018 %
- Iron (Fe):Max. 0.25 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Carbonate (CO₃):Max. 60 ppm
- Chloride (Cl):Max. 1 ppm
- Sulfate (SO₄):Max. 5 ppm
- Readily oxidizable substances:complying
- Pyridine, related substances (as Pyridine):Max. 2 ppm
- Residual solvents:complying
- Appearance of the solution:complying