1-Nitroso-2-naphthol-3,6-disulphonic acid disodium salt for spectrophotometric determination of Co, Fe, K, Ag, Ba, Ca, Co, Fe, Ni, Pb, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Synonyms: 3-Hydroxy-4-nitroso-2,7-naphthalenedisulphonic acid disodium salt, Nitroso-R salt
CAS Number:525-05-3
:
Formula:C₁₀H₅NNa₂O₈S₂
MDL Number:MFCD00003885
Molecular Weight:377.26 g/mol
Melting Pt:300 °C
EINECS:208-369-1
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity (IR):complying
- Assay:90.0 - 105.0 %