Order Entry
Singapore
ContactUsLinkComponent
Casein
Casein
Catalog # ALFAJ12840.Q1
CAS Number:  
Casein
Catalog # ALFAJ12840.Q1
CAS Number:  

Specifications

  • Pk:
    25 lb
  • CAS No.:
    9000-71-9
  • Source:
  • Species:
  • Seq:
    04
  • Environmentally Preferable:
  • Storage conditions:
    Ambient temperature
  • Form:
    Lyophilised powder
  • Protein/peptide name:
    Casein
  • Purity:
    >95%
  • Colour:
    Cream to light yellow

Specifications

About this item

Hammarsten casein is solvent extracted to remove fats from whole casein by the procedure of Hammarsten.

  • Protein: ≥95%
  • Loss on drying: ≤10,0%