Vinyltrimethoxysilane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Trimethoxy(vinyl)silane
CAS Number:2768-02-7
:
Formula:C₅H₁₂O₃Si
MDL Number:MFCD00008605
Boiling Pt:122…124 °C
Melting Pt:< –40 °C
Density:0.970
Molecular Weight:148.23 g/mol
Flash Pt:23 °C (73 °F)
EINECS:220-449-8
ADR:3 III
:GHS05
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: