Calcium bromide, ultra dry ≥99.978% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-41-5
:
Formula:Br₂Ca
MDL Number:MFCD00010902
Boiling Pt:806…812 °C
Melting Pt:730 °C
Density:3.353
Molecular Weight:199.89 g/mol
Merck Index:14,1654
:Ampuled under argon
EINECS:232-164-6
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: