Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2,6-difluorobenzoic acid
4-Chloro-2,6-difluorobenzoic acid ≥97%
4-Chloro-2,6-difluorobenzoic acid ≥97%
:
Thermo Fisher Scientific
4-Chloro-2,6-difluorobenzoic acid ≥97%
:
Thermo Fisher Scientific
:
196194-58-8
:
:
C₇H₃ClF₂O₂
:
MFCD06203570
:
192.55 g/mol
:
183…186 °C
:
GHS07
:
Ambient
Environmentally Preferable:
msds_ALFAH26556
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years