2'-Methylacetanilide ≥98%
: o-Acetotoluidide, o-Acetotoluidid, N-(o-Tolyl)acetamide, N-(2-Methylphenyl)acetamide
:120-66-1
:
:C₉H₁₁NO
:MFCD00014961
:296 °C
:109…112 °C
:1.160
:149.19 g/mol
:14,74
:204-414-4
:GHS07
:Ambient
- Environmentally Preferable: