Order Entry
Singapore
ContactUsLinkComponent
 
Acetone ≥99.5%, Laboratory Reagent
    
 
undefined
Acetone ≥99.5%, Laboratory Reagent
    

 

  • Environmentally Preferable:

 

 

  • IR spectrum:
    complying
  • Assay (GC):
    Min. 99.50 %
  • Refractive index (n 20/D):
    1.357 - 1.361
  • Non-volatile matter:
    Max. 0.002 %
  • Water (Karl Fischer):
    Max. 0.5 %
  • Explosion Limit:
    13.2 %