Nickel(II) chloride hexahydrate ≥99.95% (metals basis)
: Nickel dichloride hexahydrate
:7791-20-0
:
:NiCl₂·6H₂O
:MFCD00149809
:1.92
:237.69 g/mol
:140 °C (-H₂O)
:6.1 III
:14,6505
:231-743-0
:3288
:GHS06
:GHS08
:GHS09
:Ambient
- Environmentally Preferable: