VWR®, Slider Zip Bags
Supplier: VWR Collection
Certificates
About this item
VWR® slider zip bags, in LDPE, transparent, with blue slider. For the packing and storage of goods.
- Can be used from −10 to +65 °C
- Do not contain any ingredients of animal origin
Certifications: Manufactured under ISO 9001:2015 standard.
Packaging: 50 pcs/outer bag, 10 outer bags/case.