Acetone ≥99.5% (by GC), Technical Grade C
: 2-Propanone, DMK, Propanone, Dimethyl ketone
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:56.5 °C
:–94 °C
:0.787…0.791 kg/m³
:58.08 g/mol
:–20 °C
:Ambient
:3 II
:13,00067
:1090
:GHS02
:GHS07
- Environmentally Preferable:
- Purity (Assay G.C):99,5%
- Acidity:0,008 mEq/g
- Alkalinity:0,006 mEq/g
- Water content:0,3%
- Solubility:Soluble in water
- Miscibility:Miscible with water and most of the solvents
- Refractive index (n20/D):1,3591
- Ignition temperature:465 °C
- Vapour pressure (at 20 °C):233 hPa
- Viscosity (at 25 ˚C):0,31 mPa.s
- Dipole moment (at 20 ˚C):2,7 Debye
- Dielectric constant (at 25 ˚C):20,7
- Evaporation number (DIN 53170):2,1
- Heat evaporation (at 56 ˚C):521 KJ/Kg
- Saturation concentration (at 20 ˚C):533 g/m³
- Explosion limit (v):2,2 - 12,8 %