Optimize workflows through user-friendly handling of clearly arranged samples. Eppendorf® Tube Racks are ideal for sample processing, transport, and short- to mid-term storage of samples.
- Stackable within one rack format and also between all rack variants; saves laboratory space when not in use
- Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
- Made of polypropylene (PP) for high chemical and physical resistance
- High flexibility; different sized tubes fit in the 1.5/2.0 ml, 5.0/15.0 ml, 5.0/15/50 ml, and cryogenic tube rack
- Numbered tube positions support fast and easy orientation, and error-free handling
Benchtop racks are essential laboratory accessories for efficient processing, transport, and short-term storage of sample preparation tubes and cryogenic tubes. The comprehensive Eppendorf® Tube Rack system supports a clearly arranged and error-free handling of reaction vessels in manifold laboratory workflows. Optimized dimensions, design, and functionality result in various user benefits such as easy tube accessibility or flexible usage. The racks offer the optimal solution and perfect fit for all tubes commonly used in laboratories. Compact dimensions require minimal space.
Wavelike design: Tube rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable.
Space-saving: All racks are stackable to save lab space when not in use. Optimized space-saving dimensions due to optimized tube location-to-footprint arrangement wthout compromising the tube handling.
Material: PP
Autoclavable at 121 °C, 20 min
- Catalog No:
- EPPE0030119.860
- EPPE0030119.835
- EPPE0030119.843
- EPPE0030119.827
- EPPE0030119.851
- EPPE0030119.819
- EPPE0030119.800
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001