2-Propanol, J.T.Baker®
Catalog # BAKR3412.5000
Supplier: Avantor
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:min. 99.5 %
Specification Test Results
About this item
Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
:
:60.1 g/mol
UN:1219
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
Boiling Pt:82
Melting Pt:-89
Flash Pt:12
ADR:3 II
Merck Index:13,05228