Order Entry
Singapore
ContactUsLinkComponent
Sodium perchlorate monohydrate 98.0-102.0% Reag. USP, ACS
Sodium perchlorate monohydrate 98.0-102.0% Reag. USP, ACS
Catalog # APPC134387.1210
Supplier:  PanReac AppliChem
CAS Number:  
Sodium perchlorate monohydrate 98.0-102.0% Reag. USP, ACS
Catalog # APPC134387.1210
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White crystalline powder
  • Assay (Arg.):
    98,0 - 102,0%
  • pH of 5% solution :
    6,0-8,0
  • Insoluble matter in H₂O:
    0,005 %
  • Chloride (Cl) :
    0,003%
  • Nitrogen compounds (as N):
    0,001%
  • Phosphate (PO₄):
    0,001 %
  • Sulfate (SO₄) :
    0,002%
  • Chlorate (ClO₃) :
    0,05%
  • Heavy metals (as Pb):
    0,0005%
  • Calcium (Ca):
    0,0005%
  • Cadmium (Cd):
    0,0005%
  • Cobalt (Co):
    0,0005%
  • Copper (Cu):
    0,0005%
  • Iron (Fe):
    0,01%
  • Potassium (K):
    0,005%
  • Magnisium (Mg):
    0,0005%
  • Manganese (Mn):
    0,0005%
  • Nickel (Ni):
    0,0005%
  • Lead (Pb):
    0,0005%
  • Zinc (Zn):
    0,0005%
  • Solubility in water:
    2,090 g/l at 20 °C, hygroscopic
  • Thermal decomposition:
    Above 130 °C

Specification Test Results

About this item

Synonyms: Perchloric acid sodium salt
 :
 :140.46 g/mol
UN:1502
CAS Number:7791-07-3
Formula:NaClO₄·H₂O
MDL Number:MFCD00149164
Melting Pt:130
ADR:5.1 II
Merck Index:12,08798