Total immersion thermometers
Catalog # CGLACG-3508-N-02
Supplier: CHEMGLASS
Specifications
- Ø:6,0 - 7,0 mm
- Immersion depth:Total
- Description:White back
- Length:25 mm
- Environmentally Preferable:
- Division:1,0 °C
- Pk:1
- Range:−10...+250 °C
Specifications
About this item
Thermometer is mineral spirit filled to eliminate workplace and environmental hazards associated with mercury.
- Thermometer has a white back, a 6 to 7 mm outer diameter, and a standard taper 10/18 inner ground joint
- With 1° subdivisions
Immersion is measured from the bottom of the joint to the bottom of non-mercury bulb.