pH Papers, without color scale
These products are simple, completely impregnated indicator papers, which do not allow exact pH determinations, but pH adjustments during special chemical procedures. For this reason, no colour scale is provided with these papers.
- Long term stable and protected against external influences due to plastic packaging
- No accessories needed - always ready to use
- Quick and easy determination of the pH