Nucleic acid extraction, MPure™ Viral nucleic acids extraction kits, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
MPure™ Viral Nucleic Acids Extraction Kit is used with the MPure-12 instrument for extraction of Viral DNA or RNA from human biological specimens such as serum, plasma, and other cell-free fluids.
Specifications
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Environmentally Preferable: