Anti-beta-galactosidase Goat Polyclonal Antibody
Catalog # ICNA0856030
Supplier: MP Biomedicals
Specifications
- Antibody type:Primary
- Antigen name:beta-Galactosidase
- Antigen symbol:b-Gal
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Form:Liquid
- Storage temperature:+2 °C to +8 °C
- Size:2 mL
- Pk:2 mL
Specifications
About this item
- Liquid form
- There is no cross-reactivity to E.coli
- Ideal for IFA, cell staining (fluorescent microscopy and cell sorting), tissue staining, and blot immunostaining
Type: Primary
Antigen: beta-galactosidase
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Goat
Isotype: IgG
Reactivity: