Folin-Ciocalteu's reagent for analysis of phenols
Catalog # ICNA0219518690
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Identity Test:Passes
Specification Test Results
About this item
Synonyms: Folin & Ciocalteu’s phenol reagent
:
UN:3264
:GHS05
MDL Number:MFCD00132625
ADR:8 III