Order Entry
Singapore
ContactUsLinkComponent
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, white powder
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, white powder
Supplier:  MP Biomedicals
Certificates
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, white powder
Supplier:  MP Biomedicals

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • pH (5% aq):
    5.0-7.0
  • Optical Rotation (c=1, water):
    -26° to -33°

Specification Test Results