Order Entry
Singapore
ContactUsLinkComponent
 
α-D-Ribose-1-phosphate bis-cyclohexylammonium salt ≥96%, powder
Catalog # ICNA0215202705
Supplier:  MP Biomedicals
CAS Number:  
α-D-Ribose-1-phosphate bis-cyclohexylammonium salt ≥96%, powder
Catalog # ICNA0215202705
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

Synonyms: D-RIBOSE 1-PHOSPHATE, BISCYCLOHEXYLAMMONIUM SALT, α-D-Ribose-1-phosphate, D-Ribose 1-phosphate bis(cyclohexylammonium) salt
 :
 :428.23 g/mol
CAS Number:58459-37-3
Formula:C₁₇H₃₇N₂O₈P
MDL Number:MFCD00080934