tert-Butyl acrylate 99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Acrylic Acid tert-Butyl Ester
CAS Number:1663-39-4
:
Formula:C₇H₁₂O₂
MDL Number:MFCD00008809
Boiling Pt:121 °C
Density:0.883
:15 ppm Stabilised with 4-methoxyphenol (MEHQ)
Molecular Weight:128.17 g/mol
Melting Pt:∼–69 °C
Flash Pt:11 °C (52 °F)
Storage Temperature:Ambient
EINECS:216-768-7
UN:3272
ADR:3 II
:GHS02
:GHS07
Specifications
- Environmentally Preferable: