trans-1-Methoxy-3-trimethylsilyloxy-1,3-butadiene 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54125-02-9
:
Formula:C₈H₁₆O₂Si
MDL Number:MFCD00008498
Boiling Pt:68…69 °C (14 mmHg)
Density:0.890
Molecular Weight:172.299 g/mol
Flash Pt:45 °C (113 °F)
UN:1993
ADR:3 III
:GHS02
Specifications
- Environmentally Preferable: