Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N-Dimethylhexanamide
N,N-Dimethylhexanamide 98%
N,N-Dimethylhexanamide 98%
:
Thermo Fisher Scientific
N,N-Dimethylhexanamide 98%
:
Thermo Fisher Scientific
:
5830-30-8
:
:
CH₃(CH₂)₄CON(CH₃)₂
:
MFCD00043671
:
80 °C (1 mmHg)
:
0.890
:
143.23 g/mol
:
Ambient
:
227-406-2
:
GHS07
Environmentally Preferable:
msds_ALFAB24562
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years