Ammonium copper(II) sulphate hexahydrate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13587-26-3
:
Formula:(NH₄)₂Cu(SO₄)₂·6H₂O
MDL Number:MFCD00150440
Molecular Weight:399.83 g/mol (291.75 anhy)
:GHS06
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: