Formate, Ion chromatography standard solution, Specpure®, HCO₂- 1000 µg/ml
Molecular Formula: HCO₂Na in H₂O
Physical Form: Liquid
MDL No.: MFCD00003297
- Environmentally Preferable:
Molecular Formula: HCO₂Na in H₂O
Physical Form: Liquid
MDL No.: MFCD00003297