Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -100 mesh Type 316-L
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:Fe
MDL Number:MFCD00148499
Molecular Weight:55.845 g/mol
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: