Order Entry
Singapore
ContactUsLinkComponent
VWR® Traceable®, Hand tally counter
VWR® Traceable®, Hand tally counter
Catalog # VWRU23609-102
Supplier:  VWR Collection
undefined
VWR® Traceable®, Hand tally counter
Catalog # VWRU23609-102
Supplier:  VWR Collection
Specifications
  • Range:
    0 − 9999
  • Description:
    Manual cell counter
  • Material:
    Stainless steel
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Tally counter is designed to count blood samples, take surveys, inventory items, total drops of liquid, receive lab samples and tally biomedical events.
  • Rugged, heavy-duty chrome-finished case with hinged finger loop for grip
  • Audible click that signals the count
  • Four figures, easy-to-use
Delivery: Supplied with a loop.