Order Entry
Singapore
ContactUsLinkComponent
VWR®, Dual stage gas regulators
VWR®, Dual stage gas regulators
Supplier:  VWR Collection
Certificates
undefined
VWR®, Dual stage gas regulators
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • VWRU55850-684
  • VWRU55850-676
  • Type:
  • Hydrogen
  • Argon, nitrogen, helium
  • Max. inlet pressure:
  • 206,8 bar
  • 206,8 bar
  • Outlet pressure:
  • 0,00 - 8,61 bar
  • 0,00 - 8,61 bar
  • Flow:
  • 127,4 m³/h
  • 33,9/34,5/90,6 m³/h
Specifications