Anti-SRA1 Mouse Monoclonal Antibody [clone: 7H1G1]
Catalog # ABNOMAB10410
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Steroid receptor RNA activator 1
- Antigen symbol:SRA1
- Clonality:Monoclonal
- Clone:7H1G1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:10011
- Antigen synonyms:STRAA1|SRA|SRAP|pp7684
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human SRA1.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against recombinant SRA1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000
Type: Primary
Antigen: SRA1
Clonality: Monoclonal
Clone: 7H1G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human