Order Entry
Singapore
ContactUsLinkComponent
Anti-FFAR4 Mouse Monoclonal Antibody [clone: 2B6]
Anti-FFAR4 Mouse Monoclonal Antibody [clone: 2B6]
Catalog # ABNOMAB1725
Supplier:  Abnova
Anti-FFAR4 Mouse Monoclonal Antibody [clone: 2B6]
Catalog # ABNOMAB1725
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Free fatty acid receptor 4
  • Antigen symbol:
    FFAR4
  • Clonality:
    Monoclonal
  • Clone:
    2B6
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    338557
  • Antigen synonyms:
    GPR120|BMIQ10|GT01|PGR4|GPR129|O3FAR1
  • Storage buffer:
    PBS (0,1% proclin, 2.0% Block Ace)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human O3FAR1.
  • Size:
    25 μg
  • Pk:
    25 µg

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of O3FAR1.

Recommended Dilutions: Immunoprecipitation: 5 ?g/ml; Flow Cytometry: 1-5 ?g/ml

Type: Primary
Antigen: FFAR4
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human