Anti-NDC80 Goat Polyclonal Antibody
ABNOPAB6236
: Abnova
- Antibody type:Primary
- Antigen name:NDC80 kinetochore complex component
- Antigen symbol:NDC80
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:10403
- Antigen synonyms:HsHec1|KNTC2|TID3|hsNDC80|HEC|HEC1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-YEKKATLIKSSEE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human NDC80.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of NDC80.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: NDC80
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human