Anti-MARCH9 Rabbit Polyclonal Antibody
Catalog # ABNOPAB5435
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Septin 5
- Antigen symbol:SEPT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:92979
- Antigen synonyms:PNUTL1|CDCREL1|septin 5|H5|HCDCREL-1|CDCREL|CDCREL-1
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus of human MARCH9.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MARCH9.
Type: Primary
Antigen: MARCH9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse