Anti-FBXO6 Goat Polyclonal Antibody
Catalog # ABNOPAB6461
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:F-box protein 6
- Antigen symbol:FBXO6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:26270
- Antigen synonyms:FBG2|FBX6|Fbx6b|FBS2
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-EAAQSPYRAVVQIF
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human FBXO6.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of FBXO6.
Recommended Dilutions: ELISA: 1:8000
Type: Primary
Antigen: FBXO6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: