Anti-GPR3 Rabbit Polyclonal Antibody
ABNOPAB16749
: Abnova
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 3
- Antigen symbol:GPR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:2827
- Antigen synonyms:ACCA|ACCA orphan receptor|G-protein coupled receptor 3|GPR3
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 13 amino acids of human GPR3.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of GPR3.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: GPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat