Anti-CLASP2 Rat Monoclonal Antibody [clone: KT68]
Catalog # ABNOMAB9738
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytoplasmic Linker Associated Protein 2
- Antigen symbol:CLASP2
- Clonality:Monoclonal
- Clone:KT68
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:76499
- Antigen synonyms:mKIAA0627|C77448|CLASP2beta|8030404L10Rik|1500004F14Rik
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C for 1 month. For long term storage store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant GST fusion protein corresponding to N-terminus of mouse Clasp2.
- Purification:Protein G purification
- Pk:100 µG
Specifications
About this item
Rat monoclonal antibody raised against partial recombinant Clasp2.
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunofluorescence: 10 ?g/ml
Type: Primary
Antigen: CLASP2
Clonality: Monoclonal
Clone: KT68
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse