Anti-VCAM1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: STA]
ABNOMAB9798
: Abnova
- Antibody type:Primary
- Antigen name:Vascular cell adhesion molecule 1
- Antigen symbol:VCAM1
- Clonality:Monoclonal
- Clone:STA
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:7412
- Antigen synonyms:V-CAM 1|CD106|Vcam-1|INCAM-100|L1CAM
- Storage buffer:PBS, pH 7,4 (0,02% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
- Immunogen:Native purified VCAM1 from human DS6 T cell line.
- Purification:Size-exclusion chromatography purification
- Size:100 Reactions
- Pk:100 Tests
Mouse monoclonal antibody raised against native VCAM1.
Recommended Dilutions: Flow Cytometry: 10 ?l reagent/100 ?l of whole blood or 10^6 cells
Type: Primary
Antigen: VCAM1
Clonality: Monoclonal
Clone: STA
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human