Anti-GPR182 Rabbit Polyclonal Antibody
ABNOPAB19566
: Abnova
- Antibody type:Primary
- Antigen name:G Protein-Coupled Receptor 182
- Antigen symbol:GPR182
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:11318
- Antigen synonyms:gamrh|ADMR|hrhAMR|7TMR|G10D|AMR|AM-R
- Storage buffer:PBS.
- Storage temperature:Store at –20 °C on dry atmosphere. After reconstitution with deionized water, store at 4 °C for one month. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic protein corresponding to human GPR182.
- Purification:Protein G purification
- Size:100 μg
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic protein of GPR182.
Recommended Dilutions: ELISA: 1-2 ?g/ml; Western Blot: 2-10 ?g/ml; Immunohistochemistry: 2-10 ?g/ml
Type: Primary
Antigen: GPR182
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat