Anti-YTHDF1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB17446
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:YTH N(6)-methyladenosine RNA binding protein 1
- Antigen symbol:YTHDF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:54915
- Antigen synonyms:C20orf21
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of human YTHDF1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of YTHDF1.
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:10000
Type: Primary
Antigen: YTHDF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat