Anti-SERPINH1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB8774
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade H, member 1
- Antigen symbol:SERPINH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:871
- Antigen synonyms:Serpin H1|Cell proliferation-inducing gene 14 protein|CBP1|Arsenic-transactivated protein 3|HSP47|AsTP3|47 kDa heat shock protein|Rheumatoid arthritis-related antigen RA-A47|SERPINH2|Collagen-binding protein|PIG14|SERPINH1|CBP2|Colligin
- Storage buffer:PBS (30% glycerol, 0,5% BSA, 0,01% thimerosal)
- Storage temperature:Store at –20 °C. For long term storage store at –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to residues surrounding amino acids 407 of human SERPINH1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SERPINH1.
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunoprecipitation: 10-20 ?g/ml; Immunohistochemistry (Frozen sections): 10-20 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10-20 ?g/ml
Type: Primary
Antigen: SERPINH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chicken, Human, Mouse, Rat