Anti-SLC32A1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9637
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 32 Member A1
- Antigen symbol:Solute carrier family 32, member 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Primates,Bovine,Dog,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:83612
- Antigen synonyms:VGAT|Viaat
- Storage buffer:10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus rat Slc32a1.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of Slc32a1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: SLC32A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Primates, Rat