Anti-HVEM/TR2 Goat Polyclonal Antibody
Catalog # PIEAPA518451
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:Herpes Virus Entry Mediator
- Antigen symbol:TNFRSF14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:8764
- Antigen synonyms:member 14 (herpesvirus entry mediator)|member 14|HVEA|herpesvirus entry mediator|tumor necrosis factor receptor superfamily|HVEM|HGNC:11912|CD40-like protein precursor|ATAR|herpesvirus entry mediator A|TNFRSF14|tumor necrosis factor receptor-like gene2|TR2|LIGHTR
- Storage buffer:TBS, 0.5% BSA, 0.02% sodium azide, pH 7.3
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:0.5 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide sequence (SVQRKRQEAEGE) corresponding to the internal amino acids of TNFRSF14
- Purification:Antigen affinity chromatography
- Size:100 ug
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HVEM/TR2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human