Anti-alpha ENaC Rabbit Polyclonal Antibody
Catalog # PIEAPA1920A
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:Epithelial Sodium Channel Alpha
- Antigen symbol:SCNN1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:20276, 6337, 25122
- Antigen synonyms:alpha-epithelial sodium channel
- Storage buffer:PBS, 1mg/ml BSA, 0.05% sodium azide
- Storage temperature:-20° C, Avoid Freeze/Thaw Cycles
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic peptide corresponding to residues L(20) M K G N K R E E Q G L G P E P A A P Q Q P T(42) C of Human alpha-Enac.
- Purification:Antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: alpha ENaC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: