Anti-MCOLN2 Sheep Polyclonal Antibody
Catalog # PIEAOSM00031W
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:Mucolipin 2
- Antigen symbol:MCOLN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:whole serum
- Form:Lyophilized
- Gene ID:255231
- Antigen synonyms:mucolipin 2|TRP-ML2|TRPML2|Mucolipin-2|FLJ36691|MCOLN2|MCLN2
- Storage buffer:whole serum, no preservative
- Storage temperature:Store at 4 °C short term. For long term storage, store at –20 °C, avoiding freeze/thaw cycles.
- Shipping temperature:Ice
- Immunogen:A synthetic peptide from the c-terminal region of human Mucolipin 2 conjugated to an immunogenic carrier protein
- Size:100 µL
- Pk:100 µl
Specifications
About this item
Reconstitute with 100 ul of distilled water.
Type: Primary
Antigen: MCOLN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human