Anti-CK13 Mouse Monoclonal Antibody [clone: KS-1A3]
Catalog # PIEAMA190939
Supplier: Thermo Fisher Scientific
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 13
- Antigen symbol:CK13
- Clonality:Monoclonal
- Clone:KS-1A3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:3860
- Antigen synonyms:Keratin-13|Keratin|type I cytoskeletal 13|K13|K-13
- Storage buffer:<0.05% sodium azide
- Storage temperature:4° C, do not freeze
- Shipping temperature:Ice
- Immunogen:Cultured A431 cells from a human epidermoid carcinoma of the vulva.
- Size:1 mL
- Pk:1 mL
Specifications
About this item
MA1-90939 detects Cytokeratin 13 from human samples. MA1-90939 has been successfully used in immunohistochemistry (paraffin tissue) applications. The MA1-90939 immunogen is: Cultured A431 cells from a human epidermoid carcinoma of the vulva. Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~
Type: Primary
Antigen: CK13
Clonality: Monoclonal
Clone: KS-1A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human