Anti-CRF Mouse Monoclonal Antibody [clone: 5D5]
PIEAMA183085
- Antibody type:Primary
- Antigen name:Corticotropin releasing factor
- Antigen symbol:CRF
- Clonality:Monoclonal
- Clone:5D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:1392
- Antigen synonyms:CRF
- Storage buffer:PBS, 0.09% sodium azide
- Storage temperature:4 °C or –20 °C if preferred
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Synthetic human corticotropin factor
- Purification:Protein G
- Size:200 µg
- Pk:200 µG
Predicted to react with rat based on sequence homology. This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase. Heat-mediated antigen retrieval using a sodium citrate buffer (pH 6.0) is recommended for the staining of paraffin sections. A suggested positive control for this product is human hypothalamus. binds CRH receptors and mediates neuroendocrine stress response; may play a role in the consolidation of long term memory.
Type: Primary
Antigen: CRF
Clonality: Monoclonal
Clone: 5D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human