Anti-EYS Mouse Polyclonal Antibody
Catalog # ABNOH00346007-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Eyes Shut homolog
- Antigen symbol:EYS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:346007
- Antigen synonyms:bA74E24.1|dJ22I17.2|EGFL11|RP25|dJ1018A4.2|C6orf179|SPAM|bA166P24.2|EGFL10|bA307F22.3|C6orf180|dJ303F19.1|C6orf178
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: EYS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human