Anti-CREB3L4 Mouse Monoclonal Antibody [clone: 3E3]
Catalog # ABNOH00148327-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:cAMP responsive element binding protein 3-like 4
- Antigen symbol:CREB3L4
- Clonality:Monoclonal
- Clone:3E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- ChIP:Yes
- Gene ID:148327
- Antigen synonyms:CREB3|CREB4|AIBZIP|JAL|hJAL|ATCE1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CREB3L4
Clonality: Monoclonal
Clone: 3E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human