Anti-RAB2B Mouse Monoclonal Antibody [clone: 7E4]
ABNOH00084932-M02
: Abnova
- Antibody type:Primary
- Antigen name:RAB2B, member RAS oncogene family
- Antigen symbol:RAB2B
- Clonality:Monoclonal
- Clone:7E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:84932
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RAB2B
Clonality: Monoclonal
Clone: 7E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse