Order Entry
Singapore
ContactUsLinkComponent
 
Anti-POMK Mouse Monoclonal Antibody [clone: 6F10]
  ABNOH00084197-M03
 :  Abnova
undefined
Anti-POMK Mouse Monoclonal Antibody [clone: 6F10]
  ABNOH00084197-M03
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein-O-Mannose Kinase
  • Antigen symbol:
    POMK
  • Clonality:
    Monoclonal
  • Clone:
    6F10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • ChIP:
    Yes
  • Gene ID:
    84197
  • Antigen synonyms:
    MDDGA12|Probable Inactive Protein Kinase-Like Protein SgK196|MDDGC12|SGK196|Sugen Kinase 196|Protein Kinase-Like Protein SgK196
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: POMK
Clonality: Monoclonal
Clone: 6F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human